Bottles, narrow neck, round, LDPE, with screw caps
: BURKLE
LDPE, translucent.
- Cap absolutely tight closing
- Suitable for use with foodstuffs
- Good chemical and temperature resistance
Universal bottles for storing, packing, lab, taking samples, sampling and transport of liquids.