

2-Chloro-5-fluorophenol ≥98%
Catalog # H28567.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
CAS Number:3827-49-4
:
Formula:C₆H₄ClFO
MDL Number:MFCD00042524
:146.55 g/mol
Boiling Pt:183…185 °C
UN:3265
ADR:8,III
:GHS05
:GHS07
Storage Temperature:Ambient