

Salicylic acid ≥99.0%, AnalaR NORMAPUR® analytical reagent
Catalog # 20657.232
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:250 g
- Seq:3545
Specifications
Specification Test Results
- Assay:Min. 99.0 %
- Substances coloured by H2SO4:Passes test
- Melting point:158 - 161 °C
- Heavy metals (as Pb):Max. 10 ppm
- Ignition residue (SO4):Max. 0.05 %
- Insolubility in methanol:Max. 50 ppm
- Insolubility in NH3 4 N:Max. 50 ppm
- SO4 (Sulphate):Max. 0.025 %
- Fe (Iron):Max. 2 ppm
- Water:Max. 0.15 %
Specification Test Results
About this item
:
:138.12 g/mol
Merck Index:13,08411
Boiling Pt:336 °C (1013 hPa)
Melting Pt:157…159 °C
Density:1.443 g/cm³ (20 °C)
Flash Pt:157 °C
EINECS:200-712-3
:GHS07
:GHS05
Storage Temperature:Ambient
CAS Number:69-72-7
Formula:(HO)C₆H₄CO₂H
MDL Number:MFCD00002439