2,4-Dimethoxybenzaldehyde
Supplier: Thermo Scientific
Certificates
About this item
CAS Number:613-45-6
:
Formula:C₉H₁₀O₃
MDL Number:MFCD00003311
:166.18 g/mol
Boiling Pt:308 °C (1013 hPa)
Melting Pt:67…68 °C
Storage Temperature:Ambient
EINECS:210-342-4