
4-(Dimethylamino)benzaldehyde, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:100-10-7
:
Formula:(CH₃)₂NC₆H₄CHO
MDL Number:MFCD00003381
:149.19 g/mol
Merck Index:13,03257
Boiling Pt:176…177 °C (1013 hPa)
Melting Pt:72...75 °C (lit.)
Density:1,22 g/cm³
Flash Pt:164 °C (closed cup)
EINECS:202-819-0
:GHS07
Storage Temperature:Room Temperature
Specification Test Results
- Appearance (Colour):White to Light Yellow
- Appearance (Form):Powder
- Solubility (Colour):Colourless to Light Yellow
- Solubility (Turbidity), 50 mg/ml, EtOH:Clear
- Carbon:71.0 - 74.0%
- Nitrogen:9.1 - 9.7 %
- Purity (TLC):≥99%