Tubing, PVC, Isoflex
Supplier: KARTELL
Certificates
About this item
PVC, transparent.
- Non-toxic
- Non ageing
- Non oxiding
The tubing is marked every meter for ease of use.
Shore 'A' durometer hardness: 75 ±2.
Delivery: Supplied in 20 meter rolls packed in PE bags.