Nalgene® Packaging Bottles, Narrow Neck, Round, HDPE, with Screw Caps
Easily handle Thermo Scientific™ Nalgene™ Narrow-Mouth HDPE Bottles, conveniently packaged in rigid, lint-free shrink-wrap trays and ready to feed into filling lines for various reagents. High density polyethylene bottles include polypropylene closures and rigid, lint-free trays.
- Excellent impact and chemical resistance
- Good temperature resistance
- Food-grade
- Include opaque amber polypropylene closures
- Translucent amber bottles provide light protection with the ability to see liquid level
- Leakproof when used with Nalgene closures
Guaranteed leakproof, excellent for storage, shipping, and packaging of specialty liquid chemicals, diagnostics, biologicals, reagents, adhesives, and veterinary pharmaceuticals.
: Manufactured in compliance with ISO 13485:2003 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
- Catalog No:
- NALG322089-0008
- NALG322089-0002
- NALG322089-0001
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement