Order Entry
Sweden
ContactUsLinkComponent
4-Bromoaniline
4-Bromoaniline
Catalog # EHERC10700000
Supplier:  EHRENSTORFER
CAS Number:  
4-Bromoaniline
Catalog # EHERC10700000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,5 g
  • Seq:
    3035

Specifications

About this item

 :
Merck Index:13,01388
Boiling Pt:223 °C (1013 hPa)
Melting Pt:62…64 °C
Density:1.799 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-393-9
UN:2811
ADR:6.1,III
 :172.02 g/mol
 :GHS06
CAS Number:106-40-1
Formula:C₆H₆BrN
MDL Number:MFCD00007822