Order Entry
Sweden
Orders LinkContactUsLinkComponent
Trichloroethylene

Trichloroethylene

200 L
Catalog # SWEDTR629902
Health
Irritant
Supplier:   SWED HANDLING
Trichloroethylene
Catalog # SWEDTR629902
Supplier:  SWED HANDLING
CAS Number:  79-01-6
Trichloroethylene
Trichloroethylene

200 L

CAS Number:  79-01-6
Price
Inquire For Price
Log in for availability
1 * 200 L
Product Details & Documents
Synonyms:  1,1,2-Trichloroethene, Trichloroethene, 1,1,2-Trichloroethylene
  • CAS Number:79-01-6
  • Formula:ClCH=CCl₂
  • MDL Number:MFCD00000838
  • Molecular Weight:131.39 g/mol
  • Merck Index:12,09769
  • Boiling Pt:86.7 °C (1013 hPa)
  • Melting Pt:–73 °C
  • Density:1.46 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:201-167-4
  • UN:1710
  • ADR:6.1,III
Specifications

Specifications

  • Pk:
    200 L

Specifications