Order Entry
Sweden
ContactUsLinkComponent
Methyl ethyl ketone
Methyl ethyl ketone
Catalog # SWEDME371904
Supplier:  SWED HANDLING
   Safer Human and Environmental Health
CAS Number:  
undefined
Methyl ethyl ketone
Catalog # SWEDME371904
Supplier:  SWED HANDLING
   Safer Human and Environmental Health
CAS Number:  

Specifications

  • Pk:
    200 L
  • Seq:
    5549
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion

Specifications

About this item

 :
Merck Index:13,06097
Boiling Pt:79.6 °C (1013 hPa)
Melting Pt:–86 °C
Density:0.8054 g/cm³ (20 °C)
Flash Pt:–4 °C
Storage Temperature:Ambient
EINECS:201-159-0
UN:1193
ADR:3,II
 :72.11 g/mol
REACH:01-2119457290-43
 :GHS02
 :GHS07
CAS Number:78-93-3
Formula:C₂H₅COCH₃
MDL Number:MFCD00011648