Order Entry
Sweden
Orders LinkContactUsLinkComponent
 

Glycerine

195 L
Katalog # SWEDGL032906
   Safer Human and Environmental Health
Leverantör:   SWED HANDLING
Glycerine
Katalog # SWEDGL032906
Leverantör:  SWED HANDLING
   Safer Human and Environmental Health
CAS-nummer:  56-81-5
Glycerine
Glycerine

195 L

CAS-nummer:  56-81-5
Price
Fråga efter pris
Log in för att kolla tillgänglighet och ert pris
1 * 195 L
Product Details & Documents
Synonymer:  1,2,3-Propanetriol, Propane-1,2,3-triol, Glycerol
  • CAS-nummer:56-81-5
  • Formel:HOCH₂CH(OH)CH₂OH
  • MDL-nummer:MFCD00004722
  • Molecular Weight:92.09 g/mol
  • Merck Index:13,04497
  • Kokande Pt:290 °C (1013 hPa)
  • Smält Pt:18.6 °C
  • Densitet:1.26 g/cm³ (20 °C)
  • Flash Pt:177 °C
  • Förvaringstemperatur:Ambient
  • EINECS:200-289-5
Specifikationer

Specifikationer

  • Pk:
    195 L
  • Cat. No.:
    SWEDGL032906
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating)
  • Environmentally Preferable:
    Safer Human and Environmental Health

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.