
Acetic acid 60%
Catalog # SWEDAT861302
Supplier: SWED HANDLING
CAS Number:
Specifications
- Pk:26 kg
- Seq:4295
Specifications
About this item
:
:60.05 g/mol
Storage Temperature:Ambient
EINECS:200-580-7
UN:2790
ADR:8,II
:GHS05
CAS Number:64-19-7
Formula:H₃CCOOH
MDL Number:MFCD00036152