
Benzophenone imine ≥95%, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:1013-88-3
:
Formula:C₁₃H₁₁N
MDL Number:MFCD00001760
:181.24 g/mol
Boiling Pt:151,..153 °C/10 mmHg (lit.)
Density:1,08 g/ml (25 °C) (lit.)
:M-Clarity™ quality level = MQ100
Flash Pt:109 °C (closed cup)
:GHS07
Storage Temperature:Room Temperature
Specification Test Results
- Appearance (color):Colorless to yellow
- Appearance (form):Liquid
- Titration (nt) HClO4 0.1m:94.0 - 106.0 %
- Purity (GC area %):≥ 94.0 %
- Infrared spectrum:Conforms to structure