Ethyl pelargonate, Sigma-Aldrich®
8.20916.0250
: Merck
:
- Pack type:Glass bottle
- Pk:250 mL
- Seq:2590
- Solubility:(20 °C) insoluble
- Melting Point:-44 °C
- Molar Mass:186.29 g/mol
- Boiling Point:220 - 222 °C
- Flash Point:93 °C
- Density:0.86 g/cm3 (20 °C)
- Synonyms:Nonanoic acid ethyl ester, Ethyl pelargonate, Pelargonic acid ethyl ester
- Hill Formula:C11H22O2
- Chemical Formula:CH3(CH2)7COOC2H5
:
:M-Clarity™ quality level = MQ200
:186.29 g/mol
:204-615-7
:123-29-5
:C₁₁H₂₂O₂
:MFCD00009570