2-Ethylhexyl acrylate stabilised, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:103-11-7
:
Formula:C₁₁H₂₀O₂
MDL Number:MFCD00009495
:184.28 g/mol
:M-Clarity™ quality level = MQ200
:Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ)
EINECS:203-080-7
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:0.1 g/l (25 °C)
- Melting Point:-90 °C
- Molar Mass:184.27 g/mol
- Boiling Point:229 °C (1013 hPa)
- Vapor Pressure:0.12 hPa (20 °C)
- Flash Point:86 °C
- Explosion Limit:0.9 - 6.0 %(V)
- Density:0.887 g/cm3 (20 °C)
- Ignition Point:230 °C