Organic reference standard, 2-Nitrobiphenyl 10 µg/ml in cyclohexane, Dr Ehrenstorfer
EHERL20963200CY
:
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:86-00-0
- Description:2-Nitrobiphenyl 10 µg/ml in cyclohexane
- Formula:C₁₂H₉NO₂
- Description:2-Nitrobiphenyl