Order Entry
Sweden
ContactUsLinkComponent
Pesticide reference standard, 1,2,4-Trichlorobenzene 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Pesticide reference standard, 1,2,4-Trichlorobenzene 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL17722400CY
Supplier:  EHRENSTORFER
CAS Number:  
Pesticide reference standard, 1,2,4-Trichlorobenzene 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL17722400CY
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    120-82-1
  • Description:
    1,2,4-Trichlorobenzene 10 µg/ml in cyclohexane
  • Formula:
    C₆H₃Cl₃
  • Description:
    1,2,4-Trichlorobenzene

Specifications

About this item