2-Ethylhexyl acrylate
Catalog # EHERCA10045390
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- Seq:2070
Specifications
About this item
:
:184.28 g/mol
EINECS:203-080-7
CAS Number:103-11-7
Formula:C₁₁H₂₀O₂
MDL Number:MFCD00009495