Cholecalciferol (Vitamin D3) 99%, crystalline
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:67-97-0
:
Formula:C₂₇H₄₄O
MDL Number:MFCD00078131
:384.65 g/mol
Melting Pt:82…87 °C
Storage Temperature:Refrigerator
EINECS:200-673-2
UN:2811
ADR:6.1,II
:GHS06
:GHS08
Specification Test Results
- Appearance:White to off-white Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:82 to 87 °C
- Additional info:7-dehydrocholesterol: ≤ 0.5%
- HPLC:≥98.5 %
- Specific optical rotation:+105° to +112° (25 °C, 589 nm) (c=0.8, ethanol) (1 dm, within 30 min.)