Order Entry
Sweden
Orders LinkContactUsLinkComponent
Micro filter funnels
Supplier:   Lenz Laborglas
Micro filter funnels
Supplier:  Lenz Laborglas
Certificates
Micro filter funnels
Micro filter funnels
5 Options Available Below
Product Details & Documents
Filter funnels, made of borosilicate glass 3,3, with integrated sintered glass filter plate.
  • For filtration of very small volumes
  • Highly resistant to most chemicals such as solvents, acids and alkalis
  • Very good temperature resistance
Product Family Options
Product Information
  • Plate Ø
    Porosity
    Capacity
    Stem O-Ø
    Availability
    Price
  • 10 mm
    4
    2 ml
    6 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 520,00 kr
    520,00 kr
    1 * 1 Items
       520,00 kr
  • 10 mm
    3
    2 ml
    6 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 515,00 kr
    515,00 kr
    1 * 1 Items
       515,00 kr
  • 10 mm
    0
    2 ml
    6 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 510,00 kr
    510,00 kr
    1 * 1 Items
       510,00 kr
  • 10 mm
    1
    2 ml
    6 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 510,00 kr
    510,00 kr
    1 * 1 Items
       510,00 kr
  • 10 mm
    2
    2 ml
    6 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 510,00 kr
    510,00 kr
    1 * 1 Items
       510,00 kr