Microlab® 600 Syringe Dispensers
About this item
The Microlab® 600 dispensers are highly precise syringe pumps with a touchscreen graphical user interface designed to quickly and easily dispense liquids. This positive displacement system provides better than 99% accuracy, independent of liquid viscosity, vapour pressure and temperature. The inert fluid path minimises sample carry over and improves compatibility with harsh chemicals. There are three different dispenser models available.
- Dispense volumes from 100 µl to 50 ml
- Inert fluid path compatible with harsh chemicals
The single syringe dispenser is ideal for dedicated dispensing applications requiring a single syringe drive. The dual syringe dispenser has two syringe drives for applications such as animal dosing that require the use of more than one liquid at the same time. The dual syringe continuous dispenser is designed for time critical applications requiring repetitive dispensing.
The basic controller comes standard with a quick start run screen that provides basic diluting and dispensing functionality. The advanced controller delivers additional functions through intuitive wizards and the custom method editor. Wizard settings can be saved as favorites or custom methods on the hardware key. The basic controller can be upgraded to the advanced controller if it is not selected initially.
The standalone syringe pump offers total control with customised program methods using Visual C#®, Visual Basic®, and LabVIEW®. Commands can be deployed to any instrument on the network from anywhere in the world. The API is compatible with Windows® computers running Microsoft®. NET version 2.0 or higher.
: CE marked. CSA certified.
: The Bubble Free Prime (BFP) syringe is specifically designed for the Microlab® 600 series and the only syringe type compatible with the instrument. It has a conical plunger tip that extends through the threaded termination and into the valve. This unique design expels the air from the syringe and valve decreasing the number of priming cycles required.
: Dispensers are supplied with a Concorde hand probe (dual dispensers use the dual push button hand probe), universal valves, fill/dispense tubing, accessory holder, country specific power cord and default syringes. Standalone pumps are supplied with universal valves, country specific power cord and default syringes. To order a dispenser or standalone pump with other syringe sizes, please contact your local Avantor sales office.
7 Options Available Below
Product Details & Documents
The Microlab® 600 dispensers are highly precise syringe pumps with a touchscreen graphical user interface designed to quickly and easily dispense liquids. This positive displacement system provides better than 99% accuracy, independent of liquid viscosity, vapour pressure and temperature. The inert fluid path minimises sample carry over and improves compatibility with harsh chemicals. There are three different dispenser models available.
- Dispense volumes from 100 µl to 50 ml
- Inert fluid path compatible with harsh chemicals
The single syringe dispenser is ideal for dedicated dispensing applications requiring a single syringe drive. The dual syringe dispenser has two syringe drives for applications such as animal dosing that require the use of more than one liquid at the same time. The dual syringe continuous dispenser is designed for time critical applications requiring repetitive dispensing.
The basic controller comes standard with a quick start run screen that provides basic diluting and dispensing functionality. The advanced controller delivers additional functions through intuitive wizards and the custom method editor. Wizard settings can be saved as favorites or custom methods on the hardware key. The basic controller can be upgraded to the advanced controller if it is not selected initially.
The standalone syringe pump offers total control with customised program methods using Visual C#®, Visual Basic®, and LabVIEW®. Commands can be deployed to any instrument on the network from anywhere in the world. The API is compatible with Windows® computers running Microsoft®. NET version 2.0 or higher.
: CE marked. CSA certified.
: The Bubble Free Prime (BFP) syringe is specifically designed for the Microlab® 600 series and the only syringe type compatible with the instrument. It has a conical plunger tip that extends through the threaded termination and into the valve. This unique design expels the air from the syringe and valve decreasing the number of priming cycles required.
: Dispensers are supplied with a Concorde hand probe (dual dispensers use the dual push button hand probe), universal valves, fill/dispense tubing, accessory holder, country specific power cord and default syringes. Standalone pumps are supplied with universal valves, country specific power cord and default syringes. To order a dispenser or standalone pump with other syringe sizes, please contact your local Avantor sales office.
Product Family Options
Single syringe dispensers
Product Information
- AvailabilityPrice
- 615-0023Single syringe dispenser with basic controller, Microlab 610-DIS, one 1 ml BFP default syringeSingle syringe dispenser with basic controller, Microlab 610-DIS, one 1 ml BFP default syringeLog in for availability1 * 1 Items
- 613-1567Single syringe dispenser with advanced controller, Microlab 620-DIS, one 1 ml BFP default syringeSingle syringe dispenser with advanced controller, Microlab 620-DIS, one 1 ml BFP default syringeLog in for availability1 * 1 Items
Dual syringe dispensers
Product Information
- AvailabilityPrice
- 615-0025Dual syringe dispenser basic controller, Microlab 615-DIS, two 1 ml BFP default syringesDual syringe dispenser basic controller, Microlab 615-DIS, two 1 ml BFP default syringesLog in for availability1 * 1 Items
- 613-2970Dual syringe dispenser with advanced controller, Microlab 625-DIS, two 1 ml BFP default syringesDual syringe dispenser with advanced controller, Microlab 625-DIS, two 1 ml BFP default syringesLog in for availability1 * 1 Items
Dual syringe continuous dispensers
Product Information
Standalone syringe pumps
Product Information
Bubble Free Prime (BFP) and SaltLine syringes
Product Information
- Catalog #VolumeDescriptionAvailabilityPrice
Disposable ClickSure™ Tips for disposable tip hand probe (0,5 to 1000 µl)
Product Information
- Catalog #SterileVolumeTip stylePackedAvailabilityPrice
Accessories
Product Information
- Catalog #AvailabilityPrice
- Luer lock conversion kit
Product Information
- Catalog #AvailabilityPrice
- Dispense tubing, volume 3,14 µl/mm, custom length, 12 gauge, FEPDispense tubing, volume 3,14 µl/mm, custom length, 12 gauge, FEPHAMI1171-02Log in for availability1 * 1 Items - 1 175,00 kr
Product Information
- Catalog #AvailabilityPrice
- Dispense tubing, volume 0,79 µl/mm, custom length, 18 gauge, FEPDispense tubing, volume 0,79 µl/mm, custom length, 18 gauge, FEPHAMI1173-02Log in for availability1 * 1 Items - 1 175,00 kr
Product Information
- Catalog #AvailabilityPrice
- Screen protector, PET
Product Information
- Catalog #AvailabilityPrice
- Universal power supply, 110 - 220 VACUniversal power supply, 110 - 220 VACHAMI61092-01Log in for availability1 * 1 Items - 1 975,00 kr
Product Information
- Catalog #AvailabilityPrice
- Cable management system for disposable tip hand probeCable management system for disposable tip hand probeHAMI65160-01Log in for availability1 * 1 Items - 2 500,00 kr
Product Information
- Catalog #AvailabilityPrice
- Airshield for fresh air inside the instrumentAirshield for fresh air inside the instrumentHAMI68562-01Log in for availability1 * 1 Items - 6 900,00 kr
Product Information
- Catalog #AvailabilityPrice
- Dual push button hand probe
Product Information
- Catalog #AvailabilityPrice
- Fill tubing, volume 3,14 µl/mm, custom length, 12 gauge, FEPFill tubing, volume 3,14 µl/mm, custom length, 12 gauge, FEPHAMI1172-02Log in for availability1 * 1 Items - 960,00 kr
Product Information
- Catalog #AvailabilityPrice
- Fill tubing, volume 0,79 µl/mm, custom length, 18 gauge, FEPFill tubing, volume 0,79 µl/mm, custom length, 18 gauge, FEPHAMI1174-02Log in for availability1 * 1 Items - 960,00 kr
Product Information
- Catalog #AvailabilityPrice
- Continuous fill tubing, volume 3,14 µl/mm, 12 gauge, FEPContinuous fill tubing, volume 3,14 µl/mm, 12 gauge, FEPHAMI61491-01Log in for availability1 * 1 Items - 1 450,00 kr
Product Information
- Catalog #AvailabilityPrice
- New Microlab 600 controller, includes SD card slot, serial port, large and small USB portNew Microlab 600 controller, includes SD card slot, serial port, large and small USB portHAMI61500-05Log in for availability1 * 1 Items - 43 775,00 kr
Product Information
- Catalog #AvailabilityPrice
- Continuous fill tubing, volume 0,79 µl/mm, 18 gauge, FEPContinuous fill tubing, volume 0,79 µl/mm, 18 gauge, FEPHAMI61491-02Log in for availability1 * 1 Items - 1 300,00 kr
Product Information
- Catalog #AvailabilityPrice
- Printer kit includes printer, power source, adapter and cablePrinter kit includes printer, power source, adapter and cable613-5822Log in for availability1 * 1 Items
Product Information
- Catalog #AvailabilityPrice
- Disposable tips for 613-1625, 5 ml
Product Information
- Catalog #AvailabilityPrice
- Foot switch for ML 600 series
Product Information
- Catalog #AvailabilityPrice
- Valve plug
Product Information
- Catalog #AvailabilityPrice
- Large volume disposable tip hand probe (1 - 5 ml)Large volume disposable tip hand probe (1 - 5 ml)613-1625Log in for availability1 * 1 Items - 24 850,00 kr
Product Information
- Catalog #AvailabilityPrice
- Probe holder and tubing stand
Product Information
- Catalog #AvailabilityPrice
- Fill tubing, volume 0,96 ml, 1219 mm, 18 gauge, FEPFill tubing, volume 0,96 ml, 1219 mm, 18 gauge, FEP613-1582Log in for availability1 * 1 Items - 705,00 kr
Product Information
- Catalog #AvailabilityPrice
- Fill tubing, volume 4,57 ml, 1219 mm, 12 gauge, FEPFill tubing, volume 4,57 ml, 1219 mm, 12 gauge, FEP613-1583Log in for availability1 * 1 Items - 675,00 kr
Product Information
- Catalog #AvailabilityPrice
- Custom programmer kit (compatible with Microsoft® .NET 2.0 and higher)Custom programmer kit (compatible with Microsoft® .NET 2.0 and higher)HAMI61500-03Log in for availability1 * 1 Items - 4 650,00 kr
Product Information
- Catalog #AvailabilityPrice
- Concorde CT hand probe
Product Information
- Catalog #AvailabilityPrice
- Valve cross tube assembly
Product Information
- Catalog #AvailabilityPrice
- Left valve assembly
Product Information
- Catalog #AvailabilityPrice
- Dispense tubing, volume 1,08 ml, 1372 mm, 18 gauge, FEPDispense tubing, volume 1,08 ml, 1372 mm, 18 gauge, FEP613-1584Log in for availability1 * 1 Items - 720,00 kr
Product Information
- Catalog #AvailabilityPrice
- Dispense tubing, volume 5,15 ml, 1372 mm, 6,35 - 711,2 mm UNF, 12 gauge, FEPDispense tubing, volume 5,15 ml, 1372 mm, 6,35 - 711,2 mm UNF, 12 gauge, FEP613-1585Log in for availability1 * 1 Items - 720,00 kr
Product Information
- Catalog #AvailabilityPrice
- Right valve assembly
Product Information
- Catalog #AvailabilityPrice
- Disposable tip hand probe, 0,5 - 1000 µlDisposable tip hand probe, 0,5 - 1000 µl613-5823Log in for availability1 * 1 Items - 11 925,00 kr
Product Information
- Catalog #AvailabilityPrice
- Tubing clips
Product Information
- Catalog #AvailabilityPrice
- Basic to Advanced Controller upgrade kit, including 2 GB SD cardBasic to Advanced Controller upgrade kit, including 2 GB SD cardHAMI61500-02Log in for availability1 * 1 Items - 11 575,00 kr