
1-Decyne 98%
:764-93-2
:
:C₁₀H₁₈
:MFCD00009576
:174 °C (760 mmHg)
:–44 °C
:0.76 g/cm³
:48 °C
:212-132-8
:3,III
:138.25 g/mol
:1993
:GHS02
:Refrigerator
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4260 to 1.4280 (20°C, 589 nm)
- GC:≥97.5 %