1-Octene for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:111-66-0
:
Formula:C₈H₁₆
MDL Number:MFCD00009548
:112.22 g/mol
Merck Index:13,01770
Boiling Pt:121 °C (1013 hPa)
Melting Pt:–102 °C
Density:0.7152 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-893-7
UN:3295
ADR:3,II
:M-Clarity™ quality level = MQ200
Flash Pt:8 °C
:GHS02
:GHS08
:GHS07
:GHS09
Specification Test Results
- Solubility:(20 °C) practically insoluble
- Melting Point:-102 °C
- Molar Mass:112.21 g/mol
- Boiling Point:121 °C
- Vapor Pressure:17.3 hPa (20 °C)
- Flash Point:8 °C
- Refractive Index:1.4085 (20 °C, 589 nm)
- Explosion Limit:0.7 - 6.8 %(V)
- Density:0.72 g/cm3 (20 °C)
- Ignition Point:221 °C
- Viscosity kinematic:0.38 mm2/s (40 °C)
- Hazard Class:3
- Hazard Class Description:Flammable Liquid