Order Entry
Sweden
ContactUsLinkComponent
1-Octene
1-Octene
Catalog # EHERC15711300
Supplier:  EHRENSTORFER
CAS Number:  
1-Octene
Catalog # EHERC15711300
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :112.22 g/mol
Merck Index:13,01770
Boiling Pt:121 °C (1013 hPa)
Melting Pt:–102 °C
Density:0.7152 g/cm³ (20 °C)
Flash Pt:8 °C
Storage Temperature:Ambient
EINECS:203-893-7
UN:3295
ADR:3,II
CAS Number:111-66-0
Formula:C₈H₁₆
MDL Number:MFCD00009548