Order Entry
Sweden
ContactUsLinkComponent
Supelco® Organic Reference Standard, 1-Octadecene
Supelco® Organic Reference Standard, 1-Octadecene
Supplier:  Merck
Certificates
undefined
Supelco® Organic Reference Standard, 1-Octadecene
Supplier:  Merck

Specifications

  • CAS No.:
    112-88-9
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H304
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00009003
  • M-Clarity quality level:
    MQ100
  • Melting point:
    14 - 16 °C (lit.)
  • Formula:
    CH₃(CH₂)₁₅CH=CH₂
  • Molecular weight:
    252,48
  • Density:
    0,789 g/ml at 25 °C (lit.)
  • Flash point:
    149,00 °C - Closed cup
  • Boiling point:
    179 °C/15 mmHg (lit.); 314,4 ±5,0 °C/760 mmHg (lit.)

Specifications

Frequently Bought Together