Order Entry
Sweden
ContactUsLinkComponent
1-Octadecene for synthesis, Sigma-Aldrich®
1-Octadecene for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
1-Octadecene for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Solubility:
    (20 °C) practically insoluble
  • Melting Point:
    17 - 18 °C
  • Molar Mass:
    252.48 g/mol
  • Boiling Point:
    315 °C (1013 hPa)
  • Vapor Pressure:
    1.3 hPa (20 °C)
  • Flash Point:
    154 °C
  • Refractive Index:
    1.4439 (20 °C, 589 nm)
  • Density:
    0.79 g/cm3 (20 °C)
  • Viscosity kinematic:
    3.6 mm2/s (40 °C)

Specification Test Results

Related Information