Order Entry
Sweden
ContactUsLinkComponent
Supelco® Organic Reference Standard, 1-Hexene
Supelco® Organic Reference Standard, 1-Hexene
Supplier:  Merck
Certificates
undefined
Supelco® Organic Reference Standard, 1-Hexene
Supplier:  Merck

Specifications

  • CAS No.:
    592-41-6
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H225 - H304
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00009505
  • M-Clarity quality level:
    MQ100
  • Formula:
    CH₃(CH₂)₃CH=CH₂
  • Molecular weight:
    84,16
  • Density:
    0,678 g/ml at 25 °C (lit.)
  • Flash point:
    –25,0 °C – Closed cup
  • Boiling point:
    60 - 66 °C (lit.)

Specifications

Frequently Bought Together