Order Entry
Sweden
ContactUsLinkComponent
Organic reference standard, Tetrachloroethylene 1000 µg/ml in methanol, Dr Ehrenstorfer
Organic reference standard, Tetrachloroethylene 1000 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERY17358300ME
Supplier:  EHRENSTORFER
CAS Number:  
Organic reference standard, Tetrachloroethylene 1000 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERY17358300ME
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    127-18-4
  • Description:
    Tetrachloroethylene 1,000 µg/ml in methanol
  • Formula:
    C₂Cl₄
  • Description:
    Tetrachloroethylene

Specifications

About this item