Order Entry
Sweden
ContactUsLinkComponent
Benzyl acetate, Supelco®
Benzyl acetate, Supelco®
Katalog # 50475-5ML
Supplier:  Merck
CAS Number:  
undefined
Benzyl acetate, Supelco®
Katalog # 50475-5ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 mL
  • Seq:
    2210

Specifications

About this item

 :
 :150.18 g/mol
Merck Index:13,01125
Boiling Pt:206 °C (1013 hPa)
Melting Pt:–51 °C
Density:1.04 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
Flash Pt:95 °C
Storage Temperature:Ambient
EINECS:205-399-7
CAS Number:140-11-4
Formula:C₉H₁₀O₂
MDL Number:MFCD00008712