Order Entry
Sweden
ContactUsLinkComponent
Benzyl acetate 99+%
Benzyl acetate 99+%
Certificates
undefined
Benzyl acetate 99+%

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5010 to 1.5030 (20°C, 589 nm)
  • GC:
    ≥99.0 %

Specification Test Results

Related Information