



2,2,4-Trimethylpentane ≥99.5%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:540-84-1
:
Formula:(CH₃)₂CHCH₂C(CH₃)₃
MDL Number:MFCD00008943
:114.23 g/mol
Merck Index:13,05212
Boiling Pt:99.2 °C (1013 hPa)
Melting Pt:–107 °C
Density:0.692 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:208-759-1
UN:1262
ADR:3,II
:Filtered through a 0.2 µm filter, packaged under nitrogen.
Flash Pt:–12 °C
:GHS02
:GHS08
:GHS07
:GHS09
Specification Test Results
- Assay (GC):Min. 99.50 %
- Water:Max. 0.01 %
- Non-volatile residue:Max. 0.0005 %
- Acidity:Max. 0.0005 meq/g
- Alkalinity:Max. 0.0002 meq/g
- Transmittance (220 nm):Min. 70.0 %
- Transmittance (235 nm):Min. 80.0 %
- Transmittance (255 nm):Min. 98.0 %
- Conforms to BDH 15246:Passes test
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101) 2,5L Glass bottle for liquids DIN 45 (ZPACK13415249)
Packaging Information
spec_VWRC83630.290 spec_VWRC83630.290 spec_VWRC83630.320 spec_VWRC83630.320