Order Entry
Sweden
ContactUsLinkComponent
Sodium dodecyl sulphate (SDS) ≥95%
Sodium dodecyl sulphate (SDS) ≥95%
Leverantör:  MP Biomedicals
Certifikat
undefined
Sodium dodecyl sulphate (SDS) ≥95%
Leverantör:  MP Biomedicals

Specifikationstestresultat

  • Synonyms:
    Sodium Lauryl sulphate; sulphuric Acid, Monododecyl Ester, Sodium Salt; Dodecyl sulphate, sodium salt; SLS; SDS; Neutrazyme; Lauryl sulphate Sodium Salt; SDS Running Buffer
  • Form:
    White Powder
  • Storage:
    Store at room temperature (15 to 30 °C)
  • Aggregation Number:
    62
  • CMC:
    8,27 mM
  • pH:
    6 - 7 (1% aq soln)
  • Refractive Index:
    1,461 (Lit.)
  • Typical Working Concentration:
    10 mg SDS/mg protein

Specifikationstestresultat