Order Entry
Sweden
ContactUsLinkComponent
Glycerine 99.5+% for spectroscopy
Glycerine 99.5+% for spectroscopy
   Sustainable Options Available
Certificates
undefined
Glycerine 99.5+% for spectroscopy
   Sustainable Options Available
Specifications
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating)
Specifications
Specification Test Results
  • Appearance:
    Clear liquid
  • Water:
    ≤0.5 % (Coulometric)
  • GC:
    ≥99.50 %
  • UV:
    1 cm cell vs water
  • Color scale:
    ≤10 APHA