Order Entry
Sweden
ContactUsLinkComponent
Silver nitrate 0.01 N in 2-propanol, AVS TITRINORM
Flammable
Irritant
Silver nitrate 0.01 N in 2-propanol, AVS TITRINORM
Catalog # 85102.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Flammable
Irritant
Silver nitrate 0.01 N in 2-propanol, AVS TITRINORM
Catalog # 85102.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :169.87 g/mol
Melting Pt:-89 °C
Storage Temperature:Ambient
EINECS:231-853-9
UN:1993
ADR:3,II
 :GHS02
 :GHS07
CAS Number:7761-88-8
Formula:AgNO₃
MDL Number:MFCD00003414