Order Entry
Sweden
ContactUsLinkComponent
2-Propanol
2-Propanol
Catalog # EHERC16415200
Supplier:  EHRENSTORFER
CAS Number:  
undefined
2-Propanol
Catalog # EHERC16415200
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • Seq:
    2070

Specifications

About this item

 :
 :60.1 g/mol
Merck Index:13,05228
Boiling Pt:82 °C (1013 hPa)
Melting Pt:–89 °C
Density:0.786 g/cm³ (20 °C)
Flash Pt:12 °C
Storage Temperature:Ambient
EINECS:200-661-7
UN:1219
ADR:3,II
REACH:01-2119457558-25
 :GHS02
 :GHS07
CAS Number:67-63-0
Formula:(CH₃)₂CHOH
MDL Number:MFCD00011674