Order Entry
Sweden
ContactUsLinkComponent
1-Propanol
1-Propanol
Catalog # EHERC16415100
Supplier:  EHRENSTORFER
CAS Number:  
undefined
1-Propanol
Catalog # EHERC16415100
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • Seq:
    2070

Specifications

About this item

 :
 :60.1 g/mol
Merck Index:13,07934
Boiling Pt:97 °C (1013 hPa)
Melting Pt:–127 °C
Density:0.8053 g/cm³ (20 °C)
Flash Pt:15 °C
EINECS:200-746-9
UN:1274
ADR:3,II
 :GHS02
 :GHS05
 :GHS07
Storage Temperature:Ambient
CAS Number:71-23-8
Formula:H₃CCH₂CH₂OH
MDL Number:MFCD00002941