1-Hexadecanol, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:36653-82-4
:
Formula:H₃C(CH₂)₁₄CH₂OH
MDL Number:MFCD00004760
:242.45 g/mol
Merck Index:13,02037
Boiling Pt:305…330 °C (1013 hPa)
Melting Pt:46…52 °C
Density:0.81 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Flash Pt:>110 °C
Storage Temperature:Ambient
EINECS:253-149-0