1-Hexadecanol
Catalog # EHERC14192500
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
:242.45 g/mol
Merck Index:13,02037
Boiling Pt:305…330 °C (1013 hPa)
Melting Pt:46…52 °C
Density:0.81 g/cm³ (20 °C)
Flash Pt:>110 °C
Storage Temperature:Ambient
EINECS:253-149-0
CAS Number:36653-82-4
Formula:H₃C(CH₂)₁₄CH₂OH
MDL Number:MFCD00004760