

1-dekanol 98+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:112-30-1
:
Formula:H₃C(CH₂)₈CH₂OH
MDL Number:MFCD00004747
:158.28 g/mol
Merck Index:13,02875
Boiling Pt:220…240 °C (760 mmHg)
Melting Pt:6 °C
Density:0,829 g/cm³
Flash Pt:82 °C
Storage Temperature:Omgivande
EINECS:203-956-9
ADR:9,III
:GHS07
:GHS09
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4350 to 1.4390 (20°C, 589 nm)
- GC:≥98.0 %
- Chloride (Cl):≤3 ppm
- Sulfate (SO4):≤5 ppm
- Iron (Fe):≤1 ppm
- Heavy metals (as Pb):≤1 ppm