Order Entry
Sweden
ContactUsLinkComponent
Diethyl ether
Diethyl ether
Catalog # EHERC12606500
Supplier:  EHRENSTORFER
CAS Number:  
undefined
Diethyl ether
Catalog # EHERC12606500
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 mL
  • Seq:
    2210

Specifications

About this item

 :
 :74.12 g/mol
Merck Index:12,03852
Boiling Pt:35 °C (1013 hPa)
Melting Pt:–123 °C
Density:0.7135 g/cm³ (20 °C)
Flash Pt:–40 °C
Storage Temperature:Ambient
EINECS:200-467-2
UN:1155
ADR:3,I
REACH:01-2119535785-29
 :GHS02
 :GHS07
CAS Number:60-29-7
Formula:(CH₃CH₂)₂O
MDL Number:MFCD00011646