Order Entry
Sweden
ContactUsLinkComponent
Tributylamine
Toxic
Corrosive
Environment
Tributylamine
Catalog # EHERC17667500
Supplier:  EHRENSTORFER
CAS Number:  
Toxic
Corrosive
Environment
Tributylamine
Catalog # EHERC17667500
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :185.35 g/mol
Merck Index:13,09691
Boiling Pt:216 °C (1013 hPa)
Melting Pt:–70 °C
Density:0.7771 g/cm³ (20 °C)
Flash Pt:63 °C
EINECS:203-058-7
UN:2542
ADR:6.1,II
 :GHS06
 :GHS05
 :GHS09
Storage Temperature:Ambient
CAS Number:102-82-9
Formula:C₁₂H₂₇N
MDL Number:MFCD00009431