1-Chlorotetradecane
Catalog # EHERC11509500
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
:232.84 g/mol
Boiling Pt:295 °C (1013 hPa)
Melting Pt:2.2 °C
Density:0.859 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:219-368-0
CAS Number:2425-54-9
Formula:C₁₄H₂₉Cl
MDL Number:MFCD00000958