1,2,3-Trichloropropane
Catalog # EHERC17780000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pk:1 mL
- Seq:2070
Specifications
About this item
:
:147.43 g/mol
Boiling Pt:156 °C (1013 hPa)
Melting Pt:–14.7 °C
Density:1.38 g/cm³ (20 °C)
Flash Pt:74 °C
Storage Temperature:Ambient
EINECS:202-486-1
UN:2810
ADR:6.1,II
:GHS08
:GHS07
CAS Number:96-18-4
Formula:ClH₂CCHClCH₂Cl
MDL Number:MFCD00000946