Order Entry
Sweden
ContactUsLinkComponent
Nonadecanoic acid
Nonadecanoic acid
Catalog # EHERC15622300
Supplier:  EHRENSTORFER
CAS Number:  
Nonadecanoic acid
Catalog # EHERC15622300
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :298.51 g/mol
Boiling Pt:297 °C (1013 hPa)
Melting Pt:68…70 °C
Density:0,8468 g/cm³ (70 °C)
Storage Temperature:Ambient
EINECS:211-472-4
 :GHS07
CAS Number:646-30-0
Formula:C₁₉H₃₈O₂
MDL Number:MFCD00002754