Order Entry
Sweden
ContactUsLinkComponent
Acetic acid ≥99.0%, TraceSELECT™ Ultra for ultratrace analysis, Fluka™
Acetic acid ≥99.0%, TraceSELECT™ Ultra for ultratrace analysis, Fluka™
Certificates
undefined
Acetic acid ≥99.0%, TraceSELECT™ Ultra for ultratrace analysis, Fluka™

Specification Test Results

  • Assay:
    99.0 - 101.0 %
  • Silver (Ag):
    Max. 0.01 ppb
  • Aluminium (Al):
    Max. 0.2 ppb
  • Arsenic (As):
    Max. 0.5 ppb
  • Barium (Ba):
    Max. 0.01 ppb
  • Beryllium (Be):
    Max. 0.01 ppb
  • Bismuth (Bi):
    Max. 0.01 ppb
  • Calcium (Ca):
    Max. 1 ppb
  • Cadmium (Cd):
    Max. 0.01 ppb
  • Cerium (Ce):
    Max. 0.01 ppb
  • Cobalt (Co):
    Max. 0.01 ppb
  • Chromium (Cr):
    Max. 0.1 ppb
  • Cesium (Cs):
    Max. 0.01 ppb
  • Copper (Cu):
    Max. 0.2 ppb
  • Dysprosium (Dy):
    Max. 0.01 ppb
  • Erbium (Er):
    Max. 0.01 ppb
  • Europium (Eu):
    Max. 0.01 ppb
  • Iron (Fe):
    Max. 0.5 ppb
  • Gallium (Ga):
    Max. 0.01 ppb
  • Gadolinium (Gd):
    Max. 0.01 ppb
  • Germanium (Ge):
    Max. 0.02 ppb
  • Hafnium (Hf):
    Max. 0.01 ppb
  • Holmium (Ho):
    Max. 0.01 ppb
  • Indium (In):
    Max. 0.01 ppb
  • Potassium (K):
    Max. 0.5 ppb
  • Lanthanum (La):
    Max. 0.01 ppb
  • Lithium (Li):
    Max. 0.01 ppb
  • Lutetium (Lu):
    Max. 0.01 ppb
  • Magnesium (Mg):
    Max. 0.1 ppb
  • Manganese (Mn):
    Max. 0.02 ppb
  • Molybdenum (Mo):
    Max. 0.1 ppb
  • Sodium (Na):
    Max. 1 ppb
  • Neodymium (nd):
    Max. 0.01 ppb
  • Nickel (Ni):
    Max. 0.1 ppb
  • Lead (Pb):
    Max. 0.01 ppb
  • Praseodymium (Pr):
    Max. 0.01 ppb
  • Platinum (Pt):
    Max. 0.02 ppb
  • Rubidium (Rb):
    Max. 0.01 ppb
  • Rhenium (Re):
    Max. 0.01 ppb
  • Rhodium (Rh):
    Max. 0.01 ppb
  • Ruthenium (Ru):
    Max. 0.01 ppb
  • Antimony (Sb):
    Max. 0.05 ppb
  • Tin (Sn):
    Max. 1 ppb
  • Scandium (Sc):
    Max. 0.05 ppb
  • Selenium (Se):
    Max. 1 ppb
  • Samarium (Sm):
    Max. 0.01 ppb
  • Strontium (Sr):
    Max. 0.01 ppb
  • Terbium (Tb):
    Max. 0.01 ppb
  • Tellurium (Te):
    Max. 0.01 ppb
  • Thorium (Th):
    Max. 0.01 ppb
  • Titanium (Ti):
    Max. 0.05 ppb
  • Thallium (Tl):
    Max. 0.01 ppb
  • Thulium (Tm):
    Max. 0.01 ppb
  • Uran (U):
    Max. 0.01 ppb
  • Vanadium (V):
    Max. 0.01 ppb
  • Tungsten (W):
    Max. 0.01 ppb
  • Yttrium (Y):
    Max. 0.01 ppb
  • Ytterbium (Yb):
    Max. 0.01 ppb
  • Zinc (Zn):
    Max. 0.2 ppb
  • Zirconium (Zr):
    Max. 0.01 ppb
  • Autoignition Temperature:
    800 °F
  • Explosion Limit:
    4 %, 59 °F

Specification Test Results

Related Information