Acetic acid glacial 99-100%, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:64-19-7
:
Formula:H₃CCOOH
MDL Number:MFCD00036152
:60.05 g/mol
Storage Temperature:Ambient
EINECS:200-580-7
UN:2789
ADR:8,II
:GHS02
:GHS05
Specification Test Results
- Assay:min. 99.7 %
- Acetic anhydride ((CH3CO)2O):max. 0.01 %
- Color (APHA):max. 10
- Dilution Test:passes test
- Residue after Evaporation:max. 0.001 %
- Substances Reducing Dichromate:passes test
- Substances Reducing KMnO4:passes test
- Titrable Base (meq/g):max. 0.0004
- Aluminium (Al):max. 0.1
- Arsenic (As):max. 0.05
- Chloride (Cl):max. 1
- Copper (Cu):max. 0.1
- Iron (Fe):max. 0.2
- Nickel (Ni):max. 0.1
- Heavy Metals (as Pb):max. 0.5
- Sulfate (SO4):max. 1