Order Entry
Sweden
ContactUsLinkComponent
 
Acetic acid 99.8-100.5%, Puriss., meets analytical specification of BP, Ph. Eur., USP, FCC, Fluka™
 
undefined
Acetic acid 99.8-100.5%, Puriss., meets analytical specification of BP, Ph. Eur., USP, FCC, Fluka™
 

 

  • Appearance:
    complying
  • Assay:
    99.8 - 100.5 %
  • Congealing - Freezing Point:
    Min. 15.6 °C
  • Non-volatile matter:
    Max. 0.005 %
  • Aluminium (Al):
    Max. 0.00005 %
  • Copper (Cu):
    Max. 0.0005 %
  • Iron (Fe):
    Max. 2 ppm
  • Lead (Pb):
    Max. 0.5 ppm
  • Zinc (Zn):
    Max. 0.0005 %
  • Heavy metals (as Pb):
    Max. 2 ppm
  • Chloride (Cl):
    Max. 0.0002 %
  • Sulfate (SO₄):
    Max. 0.0005 %
  • KMnO₄ reducing matter (as HCOOH):
    Max. 0.01 %
  • KMnO₄ reducing substances:
    complying
  • K₂Cr₂O₇ reducing matter:
    complying
  • Residual solvents:
    complying
  • Identity USP:
    complying
  • Explosion Limit:
    4 %, 59 °F
  • Vapor Density:
    2.07 (vs air)
  • Refractive Index:
    n20/D 1.371 (lit.)