Order Entry
Sweden
ContactUsLinkComponent
Ethyl acetate, EMPLURA®, Supelco®
Ethyl acetate, EMPLURA®, Supelco®
Catalog # 8.22277.2500
Supplier:  Merck
   Safer Human and Environmental Health
CAS Number:  
undefined
Ethyl acetate, EMPLURA®, Supelco®
Catalog # 8.22277.2500
Supplier:  Merck
   Safer Human and Environmental Health
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    2,5 L
  • Seq:
    5096
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default and after discussion

Specifications

About this item

 :
 :88.11 g/mol
Merck Index:13,03792
Boiling Pt:77.1 °C (1013 hPa)
Melting Pt:–83 °C
Density:0.902 g/cm³ (20 °C)
Flash Pt:–4 °C
Storage Temperature:Ambient
EINECS:205-500-4
UN:1173
ADR:3,II
 :M-Clarity™ quality level = MQ200
REACH:01-2119475103-46
 :GHS02
 :GHS07
CAS Number:141-78-6
Formula:CH₃COOC₂H₅
MDL Number:MFCD00009171