Order Entry
Sweden
ContactUsLinkComponent
Ethyl acetate
Ethyl acetate
Catalog # EHERC13319000
Supplier:  EHRENSTORFER
CAS Number:  
undefined
Ethyl acetate
Catalog # EHERC13319000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • Seq:
    2070

Specifications

About this item

 :
 :88.11 g/mol
Merck Index:13,03792
Boiling Pt:77.1 °C (1013 hPa)
Melting Pt:–83 °C
Density:0.902 g/cm³ (20 °C)
Flash Pt:–4 °C
Storage Temperature:Ambient
EINECS:205-500-4
UN:1173
ADR:3,II
REACH:01-2119475103-46
 :GHS02
 :GHS07
CAS Number:141-78-6
Formula:CH₃COOC₂H₅
MDL Number:MFCD00009171