Ethyl acetate ≥99.5%, Environmental Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:141-78-6
:
Formula:CH₃COOC₂H₅
MDL Number:MFCD00009171
:88.11 g/mol
Merck Index:14,3757
Boiling Pt:77 °C
Melting Pt:–84 °C
Density:0.902
Flash Pt:–3 °C (26 °F)
Storage Temperature:Ambient
EINECS:205-500-4
UN:1173
ADR:3,II
REACH:01-2119475103-46
:GHS02
:GHS07
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion